• USA Home
  • Search Results

  • acridineorangehemizincchloridesalt369961012702311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acridine Orange hemi(zinc chloride) salt

2 Products
Synonym:   3,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt, 3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt), Basic Orange 14
Image of Acridine Orange hemi(zinc chloride) salt
CAS Number:
Linear Formula:
 C17H20ClN3 · HCl · 1/2ZnCl2
Molecular Weight:
 369.96
 Beilstein Registry Number:
 3734978
 Colour Index Number:
 46005
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Chemical Composition
Show More