• USA Home
  • Search Results

  • acroleindnphsolution2361888854011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acrolein-DNPH solution

1 Products
Synonym:   Acrolein-2,4-DNPH solution, Acrolein-2,4-dinitrophenylhydrazone solution
CAS Number:
Empirical Formula (Hill Notation):
 C9H8N4O4
Molecular Weight:
 236.18
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More