• USA Home
  • Search Results

  • acrylamide71087906111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acrylamide

9 Products
Synonym:   2-Propenamide, Acrylic acid amide
Image of Acrylamide
CAS Number:
Linear Formula:
 CH2=CHCONH2
Molecular Weight:
 71.08
 Beilstein/REAXYS Number:
 605349
 EC Number:
 MDL number:
 NACRES:
 NB.22
 PubChem Substance ID:
 eCl@ss:
 39031205
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  pH Value
  Vapor pressure
  Transition temperature
  Purity Grade
  Product Line
  Density
  Conductivity
  Bulk Density
  Autoignition temp
  Restriction
  Physical Form
  Suitability
  Melting Point
  Potency
  Special Grade
  Application(s)
  Solubility
Show More