• USA Home
  • Search Results

  • acrylamidesolution71087906111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acrylamide solution

2 Products
Image of Acrylamide solution
CAS Number:
Linear Formula:
 CH2=CHCONH2
Molecular Weight:
 71.08
 Beilstein/REAXYS Number:
 6185892
 MDL number:
 NACRES:
 NB.22
 PubChem Substance ID:
 eCl@ss:
 39031205
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
Show More