• USA Home
  • Search Results

  • actinomycind1255425076011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Actinomycin D

4 Products
Synonym:   Actinomycin C1, Actinomycin IV, Dactinomycin
Image of Actinomycin D
CAS Number:
Empirical Formula (Hill Notation):
 C62H86N12O16
Molecular Weight:
 1255.42
 Beilstein Registry Number:
 605235
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Mode of Action
  Activity Spectrum
  Physical Form
  Biological Source
  Assay
Show More