• USA Home
  • Search Results

  • activatedcharcoal1201744044011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Activated charcoal

19 Products
Synonym:   Charcoal activated
Image of Activated charcoal
CAS Number:
Empirical Formula (Hill Notation):
 C
Molecular Weight:
 12.01
 EC Number:
 MDL number:
 NACRES:
 SB.52
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Loss
  Miscellaneous Specifications
  Application(s)
  Surface Dimension
  Manufacturer designation
  Particle Dimension
  Greener Alternative Principles
Show More

Product #
Description
242276 Activated charcoal, DARCO®, −100 mesh particle size, powder, Green Alternative
05105 Activated charcoal, powder,
C9157 Activated charcoal, suitable for cell culture, suitable for plant cell culture,
161551 Activated charcoal, powder, -100 particle size (mesh), decolorizing, Green Alternative
31616 Activated charcoal, puriss. p.a., powder, Green Alternative
242268 Activated charcoal, DARCO®, 20-40 mesh particle size, granular, Green Alternative
C2764 Activated charcoal, untreated, granular, ≤5 mm, Green Alternative
242233 Activated charcoal, DARCO®, 4-12 mesh particle size, granular, Green Alternative
C4386 Activated charcoal, acid-washed with hydrochloric acid, Green Alternative
675326 Activated charcoal, DARCO® KB-G, Green Alternative
242241 Activated charcoal, DARCO®, 12-20 mesh, granular, Green Alternative
C2889 Activated charcoal, untreated, granular, 8-20 mesh, Green Alternative
C3014 Activated charcoal, untreated, granular, 20-60 mesh, Green Alternative
05112 Activated charcoal, for GC,
329428 Activated charcoal, NORIT® ROW 0.8 SUPRA, pellets, Green Alternative
292591 Activated charcoal, 4-14 mesh, granular, Norit® PK 3-5, Green Alternative
C5510 Activated charcoal, acid-washed with phosphoric and sulfuric acids, Green Alternative
80485 Activated charcoal, for the determination of AOX, 50-150 μm particle size,
C7606 Activated Charcoal, meets USP testing specifications,