• USA Home
  • Search Results

  • activatedcharcoalnorit1201744044011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Activated Charcoal Norit®

7 Products
Synonym:   Charcoal activated Norit®
Image of Activated Charcoal Norit<SUP>®</SUP>
CAS Number:
Empirical Formula (Hill Notation):
 C
Molecular Weight:
 12.01
 Beilstein/REAXYS Number:
 4360473
 EC Number:
 MDL number:
 NACRES:
 SB.52
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Application(s)
  Particle Dimension
  Miscellaneous Specifications
  Special Grade
  Composite Dimension
  Biological Source
  Loss
  Greener Alternative Principles
  Physical Form
  Quality clarifier
Show More