• USA Home
  • Search Results

  • acyano4hydroxycinnamicacid189172816641811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

α-Cyano-4-hydroxycinnamic acid

6 Products
Synonym:   α-CCA, α-CHCA, α-Cyano, 4-HCCA, ACCA
Image of α-Cyano-4-hydroxycinnamic acid
CAS Number:
Linear Formula:
 HOC6H4CH=C(CN)CO2H
Molecular Weight:
 189.17
 Beilstein/REAXYS Number:
 3271427
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More