• USA Home
  • Search Results

  • acyclodextrin972841001620311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

α-Cyclodextrin

6 Products
Synonym:   α-Schardinger dextrin, Cyclohexaamylose, Cyclomaltohexaose, alpha-Cyclodextrin
Image of α-Cyclodextrin
CAS Number:
Empirical Formula (Hill Notation):
 C36H60O30
Molecular Weight:
 972.84
 Beilstein/REAXYS Number:
 79627
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More