• USA Home
  • Search Results

  • adenine135137324511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Adenine

6 Products
Synonym:   6-Aminopurine, Vitamin B4
Image of Adenine
CAS Number:
Empirical Formula (Hill Notation):
 C5H5N5
Molecular Weight:
 135.13
 Beilstein/REAXYS Number:
 5777
 EC Number:
 MDL number:
 NACRES:
 NA.51
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Anal. Standard Form
  Product Line
  Molecular weight
  Biological Source
  Special Grade
  Titertest
  Solubility
  Color
  Assay
Show More