• USA Home
  • Search Results

  • adenosine267245861711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Adenosine

7 Products
Synonym:   9-β-D-Ribofuranosyladenine, Adenine riboside, Adenine-9-β-D-ribofuranoside
Image of Adenosine
CAS Number:
Empirical Formula (Hill Notation):
 C10H13N5O4
Molecular Weight:
 267.24
 Beilstein/REAXYS Number:
 93029
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 32160413
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Anal. Standard Form
  Physical Form
  Product Line
  Special Grade
Show More