• USA Home
  • Search Results

  • adenosine5diphosphatemonopotassiumsaltdihydrate501327269648111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Adenosine 5′-diphosphate monopotassium salt dihydrate

2 Products
Synonym:   ADP
Image of Adenosine 5′-diphosphate monopotassium salt dihydrate
CAS Number:
Empirical Formula (Hill Notation):
 C10H14KN5O10P2 · 2H2O
Molecular Weight:
 501.32
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
 eCl@ss:
 32160414
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More