• USA Home
  • Search Results

  • adglucose16bisphosphatetetracyclohexylammoniumsalthydrate73681anhydrousbasis7166213011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

α-D-Glucose 1,6-bisphosphate tetra(cyclohexylammonium) salt hydrate

1 Products
Synonym:   α-D-Glucose 1,6-diphosphate tetra(cyclohexylammonium) salt hydrate, Glc-1,6-PP.4CHA
Image of α-<SC>D</SC>-Glucose 1,6-bisphosphate tetra(cyclohexylammonium) salt hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C6H14O12P2 · 4C6H13N · xH2O
Molecular Weight:
 736.81 (anhydrous basis)
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More