• USA Home
  • Search Results

  • adipoylchloride1830311150211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Adipoyl chloride

1 Products
Synonym:   Adipyl chloride
Image of Adipoyl chloride
CAS Number:
Linear Formula:
 ClCO(CH2)4COCl
Molecular Weight:
 183.03
 Beilstein/REAXYS Number:
 507709
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
165212 Adipoyl chloride, 98%,