• USA Home
  • Search Results

  • adonitol1521548881311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Adonitol

2 Products
Synonym:   Adonite, Ribitol
Image of Adonitol
CAS Number:
Empirical Formula (Hill Notation):
 C5H12O5
Molecular Weight:
 152.15
 Beilstein/REAXYS Number:
 1720524
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Product Line
Show More