• USA Home
  • Search Results

  • adrenosterone3003938245611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Adrenosterone

1 Products
Synonym:   11-Ketoandrostenedione, 4-Androstene-3,11,17-trione, Androst-4-ene-3,11,17-trione, Reichstein’s substance G
Image of Adrenosterone
CAS Number:
Empirical Formula (Hill Notation):
 C19H24O3
Molecular Weight:
 300.39
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
284998 Adrenosterone, 98%,