• USA Home
  • Search Results

  • aflatoxinb1solution31227116265811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Aflatoxin B1 solution

4 Products
Image of Aflatoxin B<SUB>1</SUB> solution
CAS Number:
Empirical Formula (Hill Notation):
 C17H12O6
Molecular Weight:
 312.27
 Beilstein/REAXYS Number:
 1269174
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Purity Grade
  Package quantity
  Anal. Standard Form
  Special Grade
  Concentration
Show More