• USA Home
  • Search Results

  • aflatoxing1solution32827116539511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Aflatoxin G1 solution

3 Products
Image of Aflatoxin G<SUB>1</SUB> solution
CAS Number:
Empirical Formula (Hill Notation):
 C17H12O7
Molecular Weight:
 328.27
 Beilstein/REAXYS Number:
 1299768
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Special Grade
  Concentration
Show More