• USA Home
  • Search Results

  • agar12345900218011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Agar

19 Products
Synonym:   Agar-agar, Gum agar
CAS Number:
Linear Formula:
 (C12H18O9)n
 EC Number:
 MDL number:
 NACRES:
 NA.21
 eCl@ss:
 42040102
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Suitability
  Solubility
  Type
  Shelf Life
  Turbidity
  Loss
  Special Grade
  pH Value
  Melting Point
  Biological Source
  Gel Strength
  Transition temperature
  Titertest
Show More

Product #
Description
A1296 Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder,
A7002 Agar, ash 2.0-4.5%,
A7921 Agar, suitable for plant cell culture, powder,
05040 Agar, powder, for microbiology,
05038 Agar, for microbiology,
A6686 Agar, Bacteriological, microbiology tested, suitable for plant cell culture, granular,
A4550 Agar, Type A, suitable for plant cell culture,
05039 Agar, for microbiology,
A9799 Agar, High gel strength, suitable for plant cell culture,
A4800 Agar, Type M, suitable for plant cell culture,
A4675 Agar, Type E, suitable for plant cell culture,
A8678 Agar, suitable for plant cell culture,
W201201 Agar, fine powder, FCC,
A6549 Agar, Type A, ash 5-6%,
56763 Agar, high purity, low ionic content, low gel strenght,
05729 Agar, for chromogenic media,
42146 Agar, for membrane filtration, low gel strenght,
1.01615 Agar, extra pure, EMPROVE® ESSENTIAL Ph Eur,BP,
1012214 Agar, United States Pharmacopeia (USP) Reference Standard,