• USA Home
  • Search Results

  • agaricacid4165566699911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Agaric acid

1 Products
Synonym:   2-Hydroxy-1,2,3-nona­decane­tricarboxylic acid, Agaricic acid, Agaricin
Image of Agaric acid
CAS Number:
Empirical Formula (Hill Notation):
 C22H40O7
Molecular Weight:
 416.55
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
A7384 Agaric acid,