• USA Home
  • Search Results

  • agarose12345901236611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Agarose

21 Products
Image of Agarose
CAS Number:
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Gel Strength
  Product Line
  Manufacturer designation
  Loss
  Ice Shipping
  Assay
  Bulk Density
  Turbidity
  Special Grade
  Melting Point
  Application(s)
  Biological Source
  Type
  Physical Form
  Transition temperature
Show More

Product #
Description
A9539 Agarose, BioReagent, for molecular biology, low EEO,
A6013 Agarose, Type I, low EEO,
A4718 Agarose, High resolution, for molecular biology,
A0169 Agarose, Type I-A, low EEO,
A6877 Agarose, Medium EEO, for molecular biology,
05066 Agarose, High EEO, for molecular biology,
A5304 Agarose, Special High EEO,
A4679 Agarose, Low EEO, for Immunoelectrophoresis,
A2929 Agarose, For pulsed field electrophoresis running gel,
A2576 Agarose, Ultra-low Gelling Temperature, molecular biology grade,
A5030 Agarose, Ultra-low Gelling Temperature,
A0576 Agarose, Low EEO,
A7431 Agarose, BioReagent, for molecular biology, Wide range/Standard 3:1,
A9918 Agarose, Type II-A, Medium EEO,
A2790 Agarose, Wide range, for molecular biology,
A9793 Agarose, High EEO,
A7174 Agarose, High Gelling Temperature,
A3643 Agarose, Type IV, Special High EEO,
1.01236 Agarose, for analytical nucleic acid electrophoresis,
1.16802 Agarose, (low electroendoosmosis) for electrophoresis,
1.16801 Agarose, (medium electroendoosmosis) for electrophoresis,