• USA Home
  • Search Results

  • aglycerophosphatedehydrogenasefromrabbitmuscle12345907565411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

α-Glycerophosphate Dehydrogenase from rabbit muscle

2 Products
Synonym:   sn-Glycerol-3-phosphate:NAD+ 2-oxidoreductase, sn-Glycerol-3-phosphate Dehydrogenase from rabbit muscle
CAS Number:
 EC Number:
 Enzyme Commission (EC) Number:
 1.1.1.8  ( BRENDA | IUBMB )
 MDL number:
 NACRES:
 NA.54
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More