• USA Home
  • Search Results

  • ahexylcinnamaldehyde2163210186011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

α-Hexylcinnamaldehyde

4 Products
Synonym:   Hexyl cinnamic aldehyde
Image of α-Hexylcinnamaldehyde
CAS Number:
Linear Formula:
 C6H5CH=C[(CH2)5CH3]CHO
Molecular Weight:
 216.32
 Council of Europe no.:
 129
 FEMA Number:
 2569
 Flavis number:
 5.041
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More