• USA Home
  • Search Results

  • aketoglutaricacid1461032850711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

α-Ketoglutaric acid

4 Products
Synonym:   2-Oxoglutaric acid, 2-Oxopentanedioic acid, alpha-Ketoglutarate, alpha-Ketoglutaric acid
Image of α-Ketoglutaric acid
CAS Number:
Linear Formula:
 HOOCCH2CH2COCOOH
Molecular Weight:
 146.10
 Beilstein/REAXYS Number:
 1705689
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More