• USA Home
  • Search Results

  • alactosemonohydrate36031598981111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

α-Lactose monohydrate

3 Products
Synonym:   β-D-Gal-(1→4)-α-D-Glc, 4-O-β-D-Galactopyranosyl-α-D-glucose, Milk sugar
Image of α-Lactose monohydrate
CAS Number:
Empirical Formula (Hill Notation):
 C12H22O11 · H2O
Molecular Weight:
 360.31
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More