• USA Home
  • Search Results

  • alanycarb399538313001211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Alanycarb

1 Products
Image of Alanycarb
CAS Number:
Empirical Formula (Hill Notation):
 C17H25N3O4S2
Molecular Weight:
 399.53
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More