• USA Home
  • Search Results

  • albuminacetylatedfrombovineserum12345904846811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Albumin, Acetylated from bovine serum

2 Products
Synonym:   Ac-BSA, Acetyl BSA, Acetylated BSA
Image of Albumin, Acetylated from bovine serum
CAS Number:
 EC Number:
 MDL number:
 NACRES:
 NA.31
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
  Purification method
  Geographic origin
  Biological Source
Show More