• USA Home
  • Search Results

  • albuminfromchickeneggwhite12345900659111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Albumin from chicken egg white

6 Products
Synonym:   Albumin from hen egg white, Ovalbumin
CAS Number:
 EC Number:
 MDL number:
 NACRES:
 NA.25
 eCl@ss:
 42010201
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Biological Source
  Molecular weight
  Purification method
  Type
  Solubility
  Assay
  Physical Form
  Application(s)
Show More