• USA Home
  • Search Results

  • alcohol46076417511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Alcohol

2 Products
Synonym:   Ethanol, Ethyl alcohol
Image of Alcohol
CAS Number:
Linear Formula:
 CH3CH2OH
Molecular Weight:
 46.07
 Beilstein/REAXYS Number:
 1718733
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More