• USA Home
  • Search Results

  • aldicarb1902611606311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Aldicarb

2 Products
Synonym:   O-(Methylcarbamoyl)-2-methyl-2-(methylthio)propionaldehyd-oxime
Image of Aldicarb
CAS Number:
Empirical Formula (Hill Notation):
 C7H14N2O2S
Molecular Weight:
 190.26
 Beilstein/REAXYS Number:
 2209304
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Shelf Life
  Special Grade
Show More