• USA Home
  • Search Results

  • aldicarbnmethyl13cd3carbamoyl13c19527126117075511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Aldicarb-(N-methyl-13C,d3 carbamoyl-13C)

2 Products
Synonym:   (1E)-2-Methyl-2-(methylthio)-O-[(methyl-13C-d3-amino)carbonyl-13C]oxime propanal
Image of Aldicarb-(<I>N</I>-<I>methyl</I>-<SUP>13</SUP>C,d<SUB>3</SUB> <I>carbamoyl</I>-<SUP>13</SUP>C)
CAS Number:
Empirical Formula (Hill Notation):
 13C2C5D3H11N2O2S
Molecular Weight:
 195.27
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More