• USA Home
  • Search Results

  • aldicarbnmethyl13cd3carbamoyl13csulfoxide21127126117077711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Aldicarb-(N-methyl-13C,d3, carbamoyl-13C) sulfoxide

2 Products
Image of Aldicarb-(<I>N</I>-<I>methyl</I>-<SUP>13</SUP>C,d<SUB>3</SUB>, <I>carbamoyl</I>-<SUP>13</SUP>C) sulfoxide
CAS Number:
Empirical Formula (Hill Notation):
 13C2C5D3H11N2O3S
Molecular Weight:
 211.27
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Isotopic Purity
  Assay
Show More