• USA Home
  • Search Results

  • aldicarbsulfone22226164688411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Aldicarb-sulfone

2 Products
Image of Aldicarb-sulfone
CAS Number:
Empirical Formula (Hill Notation):
 C7H14N2O4S
Molecular Weight:
 222.26
 Beilstein/REAXYS Number:
 2215242
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Shelf Life
  Special Grade
Show More