• USA Home
  • Search Results

  • aldrithiol222031212703911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Aldrithiol-2

1 Products
Synonym:   2,2′-Dipyridyl disulfide, 2,2′-Dithiodipyridine
Image of Aldrithiol<SUP>™</SUP>-2
CAS Number:
Empirical Formula (Hill Notation):
 C10H8N2S2
Molecular Weight:
 220.31
 Beilstein/REAXYS Number:
 154629
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
143049 Aldrithiol-2, 98%,