• USA Home
  • Search Results

  • alfacalcidolforsystemsuitabilitya400644129456811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Alfacalcidol for system suitability A

1 Products
Synonym:   1α-Hydroxycholecalciferol, 1α-Hydroxyvitamin D3, Alfacalcidol
Image of Alfacalcidol for system suitability A
CAS Number:
Empirical Formula (Hill Notation):
 C27H44O2
Molecular Weight:
 400.64
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More