• USA Home
  • Search Results

  • alizarin240217248011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Alizarin

2 Products
Synonym:   1,2-Dihydroxyanthraquinone, Mordant Red 11
Image of Alizarin
CAS Number:
Empirical Formula (Hill Notation):
 C14H8O4
Molecular Weight:
 240.21
 Beilstein Registry Number:
 1914037
 Colour Index Number:
 58000
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Absorption
  pH Range
Show More