• USA Home
  • Search Results

  • alizarin3methyliminodiaceticacid38532395278111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Alizarin-3-methyliminodiacetic acid

1 Products
Synonym:   3,4-Dihydroxyanthraquinon-2-yl-methylimino-diacetic acid, Alizarin Complexone, Alizarin Fluorine Blue
Image of Alizarin-<WBR>3-<WBR>methyliminodiacetic acid
CAS Number:
Empirical Formula (Hill Notation):
 C19H15NO8
Molecular Weight:
 385.32
 Beilstein Registry Number:
 2190028
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More