• USA Home
  • Search Results

  • allene400646349011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Allene

1 Products
Synonym:   Propadiene
Image of Allene
CAS Number:
Linear Formula:
 H2C=C=CH2
Molecular Weight:
 40.06
 Beilstein/REAXYS Number:
 1730774
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More

Product #
Description
294985 Allene, ≥95%,