• USA Home
  • Search Results

  • allopurinolrelatedcompounde183163105519311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Allopurinol Related Compound E

2 Products
Synonym:   Ethyl 3-(formylamino)-1H-pyrazole-4-carboxylate, Ethyl 5-(formylamino)-1H-pyrazole-4-carboxylate
CAS Number:
Empirical Formula (Hill Notation):
 C7H9N3O3
Molecular Weight:
 183.16
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More