• USA Home
  • Search Results

  • alloxanmonohydrate16008224411311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Alloxan monohydrate

1 Products
Synonym:   2,4,5,6(1H,3H)-Pyrimidinetetrone, 2,4,5,6-Tetraoxypyrimidine, 5,6-Dioxyuracil
Image of Alloxan monohydrate
CAS Number:
Empirical Formula (Hill Notation):
 C4H2N2O4 · H2O
Molecular Weight:
 160.08
 Beilstein/REAXYS Number:
 4573597
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Solubility
Show More

Product #
Description
A7413 Alloxan monohydrate, 98%,