• USA Home
  • Search Results

  • allylalcohol580810718611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Allyl alcohol

3 Products
Synonym:   2-Propen-1-ol
Image of Allyl alcohol
CAS Number:
Linear Formula:
 CH2=CHCH2OH
Molecular Weight:
 58.08
 Beilstein/REAXYS Number:
 605307
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Application(s)
  Assay
Show More