• USA Home
  • Search Results

  • allylisocyanate8309147623911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Allyl isocyanate

1 Products
Image of Allyl isocyanate
CAS Number:
Linear Formula:
 CH2=CHCH2NCO
Molecular Weight:
 83.09
 Beilstein/REAXYS Number:
 506106
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
243272 Allyl isocyanate, 98%,