• USA Home
  • Search Results

  • allylsulfide1142159288111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Allyl sulfide

4 Products
Synonym:   3,3′-Thiobis-1-propene, Diallyl sulfide, NSC 20947, Thioallyl ether
Image of Allyl sulfide
CAS Number:
Linear Formula:
 (CH2=CHCH2)2S
Molecular Weight:
 114.21
 Beilstein/REAXYS Number:
 1736016
 Council of Europe no.:
 2174
 EC Number:
 FEMA Number:
 2042
 Flavis number:
 12.088
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
  Special Grade
  Application(s)
  Assay
Show More