• USA Home
  • Search Results

  • alternariol2582364138311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Alternariol

1 Products
Synonym:   3,7,9-Trihydroxy-1-methyl-6H-dibenzo[b,d]pyran-6-one, 3,7,9-Trihydroxy-1-methyl-benzo[c]chromen-6-one
Image of Alternariol
CAS Number:
Empirical Formula (Hill Notation):
 C14H10O5
Molecular Weight:
 258.23
 Beilstein/REAXYS Number:
 244839
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More