• USA Home
  • Search Results

  • alternariol9methylether272252345205311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Alternariol-9-methyl ether

1 Products
Synonym:   3,7-Dihydroxy-9-methoxy-1-methyl-6H-dibenzo[b,d]pyran-6-one, Alternariol monomethyl ether
Image of Alternariol-9-methyl ether
CAS Number:
Empirical Formula (Hill Notation):
 C15H12O5
Molecular Weight:
 272.25
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More