• USA Home
  • Search Results

  • alumina10196134428111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Alumina

1 Products
Synonym:   Alumina, Aluminum oxide
Image of Alumina
CAS Number:
Linear Formula:
 Al2O3
Molecular Weight:
 101.96
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More