• USA Home
  • Search Results

  • aluminosilicatemesostructured12345131802111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Aluminosilicate, mesostructured

1 Products
Image of Aluminosilicate, mesostructured
CAS Number:
Linear Formula:
 (SiO2)x(Al2O3)y
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Pore size
  Physical Form
  Chemical Composition
  Surface Dimension
Show More