• USA Home
  • Search Results

  • aluminumchloride13334744670011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Aluminum chloride

6 Products
Synonym:   Aluminium trichloride, Aluminum(III) chloride, Trichloroaluminum
Image of Aluminum chloride
CAS Number:
Linear Formula:
 AlCl3
Molecular Weight:
 133.34
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
 eCl@ss:
 38120507
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Solubility
  Quality clarifier
  Physical Form
  Special Grade
Show More