• USA Home
  • Search Results

  • aluminumchloridehexahydrate24143778413611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Aluminum chloride hexahydrate

2 Products
Image of Aluminum chloride hexahydrate
CAS Number:
Empirical Formula (Hill Notation):
 AlCl3 · 6H2O
Molecular Weight:
 241.43
 EC Number:
 MDL number:
 NACRES:
 NA.55
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Restriction
  Quality clarifier
  Agency/Method
  Special Grade
  Physical Form
  pH Value
  Assay
Show More