• USA Home
  • Search Results

  • aluminumhydroxidehydrate7800anhydrousbasis133044511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Aluminum hydroxide hydrate

2 Products
Image of Aluminum hydroxide hydrate
CAS Number:
Linear Formula:
 Al(OH)3 · xH2O
Molecular Weight:
 78.00 (anhydrous basis)
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More